
2-(Hydroxymethyl)-12-crown-4 ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75507-26-5
:
Formula:C₉H₁₈O₅
MDL Number:MFCD00067524
:206.24 g/mol
Boiling Pt:115 °C (0.04 mmHg)
Density:1.20
Flash Pt:>110 °C (230 °F)
EINECS:278-224-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together