
4-Cyano-4-phenylcyclohexanone 97%
:25115-74-6
:
:C₁₃H₁₃NO
:MFCD00044816
:199.24 g/mol
:114…119 °C
:Ambient
:246-631-7
:3439
:6.1,III
:GHS07
- Appearance:White to beige Crystalline solid
- Infrared spectrum:Conforms
- Melting point:114 to 119 °C
- GC:≥96.0 %
Frequently Bought Together