Order Entry
Czech Republic
ContactUsLinkComponent
N-Phenylaza-15-crown-5 for synthesis, Sigma-Aldrich®
N-Phenylaza-15-crown-5 for synthesis, Sigma-Aldrich®
Catalog # 8.13885.0005
Supplier:  Merck
CAS Number:  
N-Phenylaza-15-crown-5 for synthesis, Sigma-Aldrich®
Catalog # 8.13885.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    44 - 46 °C
  • Molar Mass:
    295.38 g/mol

Specification Test Results

About this item

 :
 :295.37 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:266-470-6
CAS Number:66750-10-5
Formula:C₁₆H₂₅NO₄
MDL Number:MFCD00040513