


cis-2,6-Dimethylmorpholine 97%
:6485-55-8
:
:C₆H₁₃NO
:MFCD00078428
:115.18 g/mol
:–85 °C
:41 °C
:229-353-0
:2734
:8,II
:GHS07
:GHS05
:GHS02
:Ambient
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4450 to 1.4470 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together