1-(2-Hydroxyethyl)-2-imidazolidinone ≥75% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL378658-250ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:130.1451 g/mol
EINECS:223-032-9
CAS Number:3699-54-5
Formula:C₅H₁₀N₂O₂
MDL Number:MFCD00037883