cis-Dicyclohexano-18-crown-6 (mixture of syn-cis and anti-cis isomers), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16069-36-6
:
Formula:C₂₀H₃₆O₆
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:240-216-4
UN:2811
Frequently Bought Together