
Dicyclohexano-18-crown-6 (mixture of isomers) for synthesis, Sigma-Aldrich®
: Merck
:16069-36-6
:
:C₂₀H₃₆O₆
:MFCD00005099
:372.51 g/mol
:6.1,III
:M-Clarity™ quality level = MQ200
:240-216-4
:2811
:GHS06
:Ambient
- Melting Point:46 - 50 °C
- Molar Mass:372.51 g/mol
- Flash Point:113 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Frequently Bought Together