4-tert-Butylcyclohexyl acetate (mixture of cis and trans isomers), Sigma-Aldrich®
Catalog # SIAL347736-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
EINECS:250-954-9
CAS Number:32210-23-4
Formula:C₁₂H₂₂O₂
MDL Number:MFCD00019354