Order Entry
Czech Republic
ContactUsLinkComponent
Sulphur trioxide pyridine complex for synthesis
Irritant
Sulphur trioxide pyridine complex for synthesis
Katalogové číslo 8.18112.0100
Supplier:  Merck
CAS Number:  
Irritant
Sulphur trioxide pyridine complex for synthesis
Katalogové číslo 8.18112.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    155 - 165 °C
  • Molar Mass:
    159.16 g/mol
  • Bulk Density:
    600 kg/m3
  • pH:
    (H2O) acidic
  • Ignition Point:
    350 °C

Specification Test Results

About this item

 :
 :159.16 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ200
EINECS:247-683-3
UN:3261
 :GHS07
Storage Temperature:Ambient
CAS Number:26412-87-3
Formula:C₅H₅NO₃S
MDL Number:MFCD00012437