4'-Aminobenzo-15-crown-5 (4'-aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane), Sigma-Aldrich®
Catalog # SIAL393428-500MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:283.3202 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:60835-71-4
Formula:C₁₄H₂₁NO₅
MDL Number:MFCD00068686