Order Entry
Czech Republic
ContactUsLinkComponent
1,2-Phenylene phosphorochloridite ≥97%
1,2-Phenylene phosphorochloridite ≥97%
Catalog # ABCRB22712.0100
Supplier:  abcr
CAS Number:  
1,2-Phenylene phosphorochloridite ≥97%
Catalog # ABCRB22712.0100
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
Melting Pt:∼30 °C
Flash Pt:>110 °C (230 °F)
EINECS:216-690-3
UN:3261
ADR:8,II
CAS Number:1641-40-3
Formula:C₆H₄ClO₂P
MDL Number:MFCD00005853