

O-Ethylhydroxylamine hydrochloride 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3332-29-4
:
Formula:C₂H₈ClNO
MDL Number:MFCD00012956
Melting Pt:130…133 °C
Storage Temperature:Ambient
EINECS:222-060-9
ADR:8,II
:97.54 g/mol
:GHS07
:GHS05
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Titration Argentometric:≥99.0 %
- HPLC:≥98.5 %
Specification Test Results
Frequently Bought Together