Order Entry
Czech Republic
ContactUsLinkComponent
3-(Trifluoromethyl)cyclohexanone ≥97%
Flammable
Irritant
3-(Trifluoromethyl)cyclohexanone ≥97%
Catalog # H34087.03
CAS Number:  
Flammable
Irritant
3-(Trifluoromethyl)cyclohexanone ≥97%
Catalog # H34087.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :166.14 g/mol
Boiling Pt:82 °C (29 mmHg)
Storage Temperature:Ambient
UN:1224
ADR:3,III
 :GHS02
 :GHS07
CAS Number:585-36-4
Formula:C₇H₉F₃O
MDL Number:MFCD00034662