
Ethyl-4-oxocyclohexanecarboxylate 97%
:17159-79-4
:
:C₉H₁₄O₃
:MFCD00013285
:170.21 g/mol
:150…152 °C (40 mmHg)
:1.076 g/cm³
:110 °C
:GHS07
:Ambient
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4580 to 1.4610 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together