Methyl-2-oxocyclohexanecarboxylate 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:41302-34-5
:
Formula:C₈H₁₂O₃
MDL Number:MFCD00077699
:156.1791 g/mol
Boiling Pt:50 °C (.2 mmHg)
Density:1.109 g/cm³
Flash Pt:85 °C
Storage Temperature:Ambient
EINECS:255-306-9
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥88 % may occur as tautomers
Specification Test Results
Frequently Bought Together