Order Entry
Czech Republic
ContactUsLinkComponent
6,6'-Di-tert-butyl-4,4'-thiodi-m-cresol for synthesis, Sigma-Aldrich®
Irritant
6,6'-Di-tert-butyl-4,4'-thiodi-m-cresol for synthesis, Sigma-Aldrich®
Catalog # 8.18449.0100
Supplier:  Merck
CAS Number:  
Irritant
6,6'-Di-tert-butyl-4,4'-thiodi-m-cresol for synthesis, Sigma-Aldrich®
Catalog # 8.18449.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    158 - 161 °C
  • Molar Mass:
    358.54 g/mol
  • Flash Point:
    215 °C
  • Ignition Point:
    402 °C

Specification Test Results

About this item

 :
 :358.5374 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:202-525-2
 :GHS07
Storage Temperature:Ambient
CAS Number:96-69-5
Formula:C₂₂H₃₀O₂S
MDL Number:MFCD00026287