N-Propyl-1,3-propanediamine, Sigma-Aldrich®
Catalog # SIAL308153-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:245-869-9
UN:2734
CAS Number:23764-31-0
Formula:C₆H₁₆N₂