Order Entry
Czech Republic
ContactUsLinkComponent
N-Methyl-1,3-propanediamine, Sigma-Aldrich®
N-Methyl-1,3-propanediamine, Sigma-Aldrich®
Catalog # SIAL65690-250ML
Supplier:  Merck
CAS Number:  
N-Methyl-1,3-propanediamine, Sigma-Aldrich®
Catalog # SIAL65690-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :88.15 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:228-544-6
CAS Number:6291-84-5
Formula:C₄H₁₂N₂
MDL Number:MFCD00008209