
Tris[2-(2-methoxyethoxy)ethyl]amine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:70384-51-9
:
Formula:C₁₅H₃₃NO₆
MDL Number:MFCD00010748
:323.43 g/mol
Boiling Pt:163…171 °C (1 mmHg)
Melting Pt:< –100 °C
Density:1.011
Flash Pt:162 °C (323 °F)
EINECS:274-590-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together