Order Entry
Czech Republic
ContactUsLinkComponent
Tris[2-(2-methoxyethoxy)ethyl]amine 95%
Corrosive
Tris[2-(2-methoxyethoxy)ethyl]amine 95%
Certificates
undefined
Corrosive
Tris[2-(2-methoxyethoxy)ethyl]amine 95%

Specification Test Results

  • Appearance:
    Clear light yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4466 to 1.4486 (20°C, 589 nm)
  • GC:
    ≥94 %
  • Titration with HClO4:
    ≥94 %

Specification Test Results

Related Information

Frequently Bought Together