
Methyl malonyl chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37517-81-0
:
Formula:C₄H₅ClO₃
MDL Number:MFCD00013657
:136.53 g/mol
Boiling Pt:57…59 °C (12 mmHg)
Flash Pt:80 °C
Storage Temperature:Refrigerator
EINECS:253-540-6
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4310 to 1.4350 (20°C, 589 nm)
- Titration RCOCl:≥96.0 %
Specification Test Results
Frequently Bought Together