


Ethyl glyoxylate ∼50% in toluene
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:924-44-7
:
Formula:C₄H₆O₃
MDL Number:MFCD00044009
:102.09 g/mol
Boiling Pt:110 °C
Density:1.003
Flash Pt:7 °C (44 °F)
EINECS:213-105-3
UN:1993
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together