
Decyltrimethylammonium bromide 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2082-84-0
:
Formula:C₁₃H₃₀BrN
MDL Number:MFCD00041973
Melting Pt:243 °C
:280.30 g/mol
EINECS:218-219-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige Powder and/or chunks
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 %
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together