tert-Butyl ethyl malonate 97%
Catalog # ACRO339400250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Clear colorless to pale yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- NMR:consistent with structure
Specification Test Results
About this item
:
:188.22 g/mol
Boiling Pt:106…110 °C (760 mmHg)
Density:1.001 g/cm³
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:251-269-8
CAS Number:32864-38-3
Formula:C₉H₁₆O₄
MDL Number:MFCD00009193