1,4,7-Triazacyclononane trihydrochloride, Sigma-Aldrich®
Catalog # SIAL329495-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:238.5900 g/mol
CAS Number:58966-93-1
Formula:C₆H₁₈Cl₃N₃
MDL Number:MFCD00074887