Methyltributylammonium chloride ≥75% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:56375-79-2
:
Formula:C₁₃H₃₀ClN
MDL Number:MFCD00011847
:235.84 g/mol
:M-Clarity™ quality level = MQ200
EINECS:260-135-8
Frequently Bought Together