Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
p-Phenylenediamine-[ring-D4]
p-Phenylenediamine-[ring-D₄] (98% D), Sigma-Aldrich®
p-Phenylenediamine-[ring-D₄] (98% D), Sigma-Aldrich®
p-Phenylenediamine-[ring-D₄] (98% D), Sigma-Aldrich®
SIAL487473-1G
:
Merck
:
p-Phenylenediamine-[ring-D₄] (98% D), Sigma-Aldrich®
SIAL487473-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
6.1,III
:
M-Clarity™ quality level = MQ200
:
1673
:
GHS06
:
GHS09
:
Ambient
:
153200-72-7
:
C₆H₄D₄N₂