
Bis(pentamethylcyclopentadienyl)manganese(II) ≥98%
Catalog # 38605.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:325.39 g/mol
Melting Pt:230 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:67506-86-9
Formula:C₂₀H₃₀Mn
MDL Number:MFCD00058844