1,3-Phenylene diisocyanate, Sigma-Aldrich®
Catalog # SIAL308234-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
ADR:6.1,II
:M-Clarity™ quality level = MQ100
EINECS:204-637-7
UN:2811
CAS Number:123-61-5
Formula:C₈H₄N₂O₂
MDL Number:MFCD00002018