
3-(Methylthio)propionic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:646-01-5
:
Formula:C₄H₈O₂S
MDL Number:MFCD00059635
:120.17 g/mol
Boiling Pt:244…247 °C
Melting Pt:∼ 16 °C
Density:1.16
Flash Pt:104 °C (219 °F)
Storage Temperature:Ambient
EINECS:211-460-9
UN:3265
ADR:8,III
:GHS05
Related Information
Frequently Bought Together