Sodium deuteroxide 40% in D₂O (99% D) (Isotopic), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:14014-06-3
:
Formula:NaOD
MDL Number:MFCD00037669
:41.01 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:237-825-2
UN:1824
Frequently Bought Together