Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Sodium deuteroxide 40% (w/w) in D₂O for NMR spectroscopy (D₂O 99+ atom%D)
 
undefined
 
Sodium deuteroxide 40% (w/w) in D₂O for NMR spectroscopy (D₂O 99+ atom%D)

 

  • Appearance:
    Clear colorless Solution
  • D-Enrichment:
    ≥99.0 %
  • Concentration:
    40 to 42 % w/w
  • HPLC:
    ≥95.0 %

 

 

Frequently Bought Together