Diethyl-tert-butylmalonate, Sigma-Aldrich®
Catalog # SIAL278041-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:216.28 g/mol
:M-Clarity™ quality level = MQ100
EINECS:212-067-5
CAS Number:759-24-0
Formula:C₁₁H₂₀O₄
MDL Number:MFCD00009152