Order Entry
Czech Republic
ContactUsLinkComponent
Potassium thiocyanate 0.1 M KSCN volumetric solution, Fluka™
Potassium thiocyanate 0.1 M KSCN volumetric solution, Fluka™
Catalog # 35194-1L
CAS Number:  
Potassium thiocyanate 0.1 M KSCN volumetric solution, Fluka™
Catalog # 35194-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001

Specification Test Results

About this item

 :
Density:1.886 g/cm³
 :97.18 g/mol
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413