Dropping bottles, glass
Made of amber glass.
- Good chemical and physical properties
- For use in applications that do not involve high thermal stresses
- Amber coloured to protect light sensitive substances
Dropping bottles without cap, with thread DIN 18.
: Pipette kit and screw cap must be ordered seperately.