Glove liners, Perfect Poly® PU First Liner White
Supplier: HONEYWELL SAFETY
Certificates
About this item
50% polyamide, 50% polyester knitted glove for fine handling or for use as an underglove.
- Knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- Elasticated wrist
- Non pilling, the polyamide is suitable for precision work
- White colour enhances the glove's hygiene and cleanliness
Certifications: Cat. I
Specifications
- Catalog No:
- DALL2132200/10
- DALL2132200/8
- DALL2132200/7
- DALL2132200/9
- Model:
- PU First Liner White
- PU First Liner White
- PU First Liner White
- PU First Liner White
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Gauge:
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist