Calcein indicator, for metal titration, Supelco®
Catalog # 1.02315.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance of solution:conforms
- Loss on drying (110 °C):≤ 7.0 %
- Suitability as indicator (for metal titration):conforms
Specification Test Results
About this item
:
:622.53 g/mol
:M-Clarity™ quality level = MQ200
EINECS:215-957-1
CAS Number:1461-15-0
:154071-48-4
Formula:C₃₀H₂₆N₂O₁₃
MDL Number:MFCD00005049