Phthalhydrazide, Sigma-Aldrich®
Catalog # SIALP38803-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:162.15 g/mol
:M-Clarity™ quality level = MQ100
EINECS:215-893-4
CAS Number:1445-69-8
Formula:C₈H₆N₂O₂
MDL Number:MFCD00006888