3-Nitrophthalhydrazide, Sigma-Aldrich®
Catalog # SIAL278084-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:207.1430 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:3682-15-3
Formula:C₈H₅N₃O₄
MDL Number:MFCD00006889