1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate, Sigma-Aldrich®
Catalog # SIAL660035-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:424.33 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:286014-42-4
Formula:C₂₃H₃₃BF₄N₂
MDL Number:MFCD04973311