Pyridoxal-5'-phosphate monohydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:41468-25-1
:
Formula:C₈H₁₀NO₆P·1H₂O
MDL Number:MFCD00149414
:265.17 g/mol (247.15 anhy)
Merck Index:14,7979
Melting Pt:142 °C
Storage Temperature:Freezer
EINECS:200-208-3
Related Information
Frequently Bought Together