
2-(Chloromethyl)quinoline hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3747-74-8
:
Formula:C₁₀H₉Cl₂N
MDL Number:MFCD00012734
:214.10 g/mol
Melting Pt:183…187 °C
EINECS:223-145-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Cream to light yellow Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 %
- Solubility:(5 % in water) Clear
- Titration with HClO4:≥96.0 %
- Iron (Fe):≤20 ppm
- Potassium (K):≤100 ppm
- Sodium (Na):≤1000 ppm
- Heavy metals (ICP-OES):≤0.002 %
- Sulfur compounds (as SO4):≤0.2 %
- Insoluble matter:≤0.01 % (in dilute HCl)
Specification Test Results
Frequently Bought Together