3-(2-Benzothiazolyl)-7-(diethylamino)coumarin, Sigma-Aldrich®
Catalog # SIAL546283-100MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:350.4341 g/mol
:M-Clarity™ quality level = MQ100
EINECS:253-830-2
CAS Number:38215-36-0
Formula:C₂₀H₁₈N₂O₂S
MDL Number:MFCD00041869