Order Entry
Czech Republic
ContactUsLinkComponent
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine, Sigma-Aldrich®
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine, Sigma-Aldrich®
Catalog # SIAL675806-100MG
Supplier:  Merck
CAS Number:  
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine, Sigma-Aldrich®
Catalog # SIAL675806-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :644.64 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:221012-82-4
Formula:C₃₈H₃₄N₂O₄P₂
MDL Number:MFCD04038734