(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:442905-33-1
:
Formula:C₄₆H₅₀N₂O₄P₂
MDL Number:MFCD04974235
:756.85 g/mol
Melting Pt:190…194 °C
:Packaged under argon.
Storage Temperature:Ambient
Related Information
Frequently Bought Together