1,3,2-Benzodioxaborole (catecholborane) 1 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:274-07-7
:
Formula:C₆H₅BO₂
MDL Number:MFCD00005846
:119.92 g/mol
:M-Clarity™ quality level = MQ200
EINECS:205-991-5
Frequently Bought Together