



Propanephosphonic acid cyclic anhydride 50% in DMF, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:68957-94-8
:
Formula:C₉H₂₁O₆P₃
MDL Number:MFCD00006583
:318.19 g/mol
Storage Temperature:Ambient
EINECS:422-210-5
UN:3286
ADR:3,II
:GHS02
:GHS05
:GHS08
:GHS07
Frequently Bought Together