Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
 
Propanephosphonic acid cyclic anhydride 50% (w/w) in DMF
 
undefined
 
 
 
 
Propanephosphonic acid cyclic anhydride 50% (w/w) in DMF

 

  • Appearance:
    Clear yellow to brown Solution
  • Refractive index:
    1.4500 to 1.4540 (20°C, 589 nm)
  • Assay:
    ≥50 % w/w Solids

 

 

Frequently Bought Together