



Piperidine 20% in DMF, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-89-4
:
Formula:C₅H₁₁N
MDL Number:MFCD00005979
:85.15 g/mol
ADR:3,II
:M-Clarity™ quality level = MQ100
EINECS:203-813-0
UN:3286
:GHS02
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together