Order Entry
Czech Republic
ContactUsLinkComponent
Fluoranthene-[D₁₀] (98% D), Sigma-Aldrich®
Irritant
Environment
Fluoranthene-[D₁₀] (98% D), Sigma-Aldrich®
Catalog # SIAL456292-100MG
Supplier:  Merck
CAS Number:  
Irritant
Environment
Fluoranthene-[D₁₀] (98% D), Sigma-Aldrich®
Catalog # SIAL456292-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
 :GHS07
 :GHS09
CAS Number:93951-69-0
Formula:C₁₆D₁₀